7-Chloro-6-fluoro-1-methoxyisoquinoline

Lieferant: Apollo Scientific

PC200044-1G PC200044-500MG
APOSPC200044-1GEA 538 EUR
APOSPC200044-1G APOSPC200044-500MG
7-Chloro-6-fluoro-1-methoxyisoquinoline
7-Chloro-6-fluoro-1-methoxyisoquinoline
Formel: C₁₀H₇ClFNO
MDL: MFCD18374925
CAS-Nummer: 1202006-81-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR