5-Bromo-6-(trifluoromethyl)-1H-indole-3-carbaldehyde
Lieferant: Apollo Scientific
PC200047-1G
PC200047-500MG
APOSPC200047-1GEA
634
EUR
APOSPC200047-1G
APOSPC200047-500MG
5-Bromo-6-(trifluoromethyl)-1H-indole-3-carbaldehyde
5-Bromo-6-(trifluoromethyl)-1H-indole-3-carbaldehyde
Formel:
C₁₀H₅BrF₃NO Molecular Weight: 292.052 g/mol Schmelzpunkt: 272…275 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD23136030 CAS-Nummer: 1423757-77-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...