2-Fluoro-5-[(4-oxo-3,4-dihydrophthalazin-1-yl)methyl]benzoic acid 99%

Lieferant: Apollo Scientific

PC200051-100G PC200051-25G PC200051-5G
APOSPC200051-100GEA 380 EUR
APOSPC200051-100G APOSPC200051-25G APOSPC200051-5G
2-Fluoro-5-[(4-oxo-3,4-dihydrophthalazin-1-yl)methyl]benzoic acid 99%
2-Fluoro-5-[(4-oxo-3,4-dihydrophthalazin-1-yl)methyl]benzoic acid
Formel: C₁₆H₁₁FN₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD14636678
CAS-Nummer: 763114-26-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR