Ethyl-4,6-dihydroxy-2-(trifluoromethyl)nicotinate

Lieferant: Apollo Scientific

PC200071-1G PC200071-500MG PC200071-5G
APOSPC200071-1GEA 270 EUR
APOSPC200071-1G APOSPC200071-500MG APOSPC200071-5G
Ethyl-4,6-dihydroxy-2-(trifluoromethyl)nicotinate
Ethyl-4,6-dihydroxy-2-(trifluoromethyl)nicotinate
Formel: C₉H₈F₃NO₄
Molecular Weight: 251.16 g/mol
Schmelzpunkt: 217…219 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09881353
CAS-Nummer: 103900-77-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR