2-Chloro-3-iodo-4-(trifluoromethyl)pyridine
Lieferant: Apollo Scientific
|
Achtung
|
PC200082-1G
PC200082-500MG
PC200082-5G
APOSPC200082-1GEA
0
EUR
APOSPC200082-1G
APOSPC200082-500MG
APOSPC200082-5G
2-Chloro-3-iodo-4-(trifluoromethyl)pyridine
2-Chloro-3-iodo-4-(trifluoromethyl)pyridine
Formel:
C₆H₂ClF₃IN Schmelzpunkt: 55…56 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD12827836 CAS-Nummer: 1186405-09-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...