(1S)-1-[4-(Trifluoromethyl)phenyl]prop-2-en-1-amine hydrochloride

Lieferant: Apollo Scientific

PC200088-500MG
APOSPC200088-500MGEA 661 EUR
APOSPC200088-500MG
(1S)-1-[4-(Trifluoromethyl)phenyl]prop-2-en-1-amine hydrochloride
(1S)-1-[4-(Trifluoromethyl)phenyl]prop-2-en-1-amine hydrochloride
Formel: C₁₀H₁₀F₃N·ClH
Molecular Weight: 237.649 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12068559
CAS-Nummer: 1391528-41-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR