3-Methyl-4-(trifluoromethyl)-sydnone

Lieferant: Apollo Scientific

PC200096-500MG
APOSPC200096-500MGEA 539 EUR
APOSPC200096-500MG
3-Methyl-4-(trifluoromethyl)-sydnone
3-Methyl-4-(trifluoromethyl)-sydnone
Formel: C₄H₃F₃N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD25948961
CAS-Nummer: 1067233-79-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR