1-(3,5-Difluorophenyl)ethanol 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3,5-Difluoro-alpha-methylbenzyl alcohol
PC2001-1G
APOSPC2001-1GEA
125
EUR
APOSPC2001-1G
1-(3,5-Difluorophenyl)ethanol 97%
1-(3,5-Difluorophenyl)ethanol
Formel:
C₈H₈F₂O Molecular Weight: 158,15 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04038285 CAS-Nummer: 872181-59-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...