N-Phenyl-2-(trifluoromethyl)pyridin-4-amine

Lieferant: Apollo Scientific

PC200101-1G PC200101-500MG PC200101-5G
APOSPC200101-1GEA 671 EUR
APOSPC200101-1G APOSPC200101-500MG APOSPC200101-5G
N-Phenyl-2-(trifluoromethyl)pyridin-4-amine
N-Phenyl-2-(trifluoromethyl)pyridin-4-amine
Formel: C₁₂H₉F₃N₂
Schmelzpunkt: 135…137 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD13190612
CAS-Nummer: 234112-18-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR