Potassium trifluoro(3-phenyl-1H-pyrazol-5-yl)borate

Lieferant: Apollo Scientific
Achtung

PC200109-500MG
APOSPC200109-500MGEA 626 EUR
APOSPC200109-500MG
Potassium trifluoro(3-phenyl-1H-pyrazol-5-yl)borate
Potassium trifluoro(3-phenyl-1H-pyrazol-5-yl)borate
Formel: C₉H₇BF₃N₂·K
Molecular Weight: 250,07 g/mol
Schmelzpunkt: 180 °C
Lagertemperatur: Kühlschrank
MDL: MFCD25948965
CAS-Nummer: 1402242-81-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR