Potassium trifluoro(3-(4-methoxyphenyl)-1H-pyrazol-5-yl)borate

Lieferant: Apollo Scientific

PC200110-500MG
APOSPC200110-500MGEA 626 EUR
APOSPC200110-500MG
Potassium trifluoro(3-(4-methoxyphenyl)-1H-pyrazol-5-yl)borate
Potassium trifluoro(3-(4-methoxyphenyl)-1H-pyrazol-5-yl)borate
Formel: C₁₀H₉BF₃N₂O·K
Molecular Weight: 280,096 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD25948966
CAS-Nummer: 1402242-82-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR