4-Fluoro-7-nitro-1H-indazole
Lieferant: Apollo Scientific
|
Achtung
|
PC200113-1G
APOSPC200113-1GEA
232
EUR
APOSPC200113-1G
4-Fluoro-7-nitro-1H-indazole
4-Fluoro-7-nitro-1H-indazole
Formel:
C₇H₄FN₃O₂ Schmelzpunkt: 190…192 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03787365 CAS-Nummer: 866144-02-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...