Potassium (3-(tert-butyl)-1-methyl-1H-pyrazol-5-yl)trifluoroborate

Lieferant: Apollo Scientific

PC200115-500MG
APOSPC200115-500MGEA 698 EUR
APOSPC200115-500MG
Potassium (3-(tert-butyl)-1-methyl-1H-pyrazol-5-yl)trifluoroborate
Potassium (3-(tert-butyl)-1-methyl-1H-pyrazol-5-yl)trifluoroborate
Formel: C₈H₁₃BF₃N₂·K
Molecular Weight: 244,107 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD25948968
CAS-Nummer: 1402242-79-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR