Methyl 3-bromo-5-(((trifluoromethyl)sulphonyl)oxy)-4-(trimethylsilyl)benzoate
Lieferant: Apollo Scientific
PC200118-100MG
APOSPC200118-100MGEA
205
EUR
APOSPC200118-100MG
Methyl 3-bromo-5-(((trifluoromethyl)sulphonyl)oxy)-4-(trimethylsilyl)benzoate
Methyl 3-bromo-5-(((trifluoromethyl)sulphonyl)oxy)-4-(trimethylsilyl)benzoate
Formel:
C₁₂H₁₄BrF₃O₅SSi Molecular Weight: 435,286 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD25948969 CAS-Nummer: 1242069-32-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...