5-Chloro-3-(trifluoromethyl)-2-pyridine carboxylic acid

Lieferant: Apollo Scientific

PC200119-500MG
APOSPC200119-500MGEA 764 EUR
APOSPC200119-500MG
5-Chloro-3-(trifluoromethyl)-2-pyridine carboxylic acid
5-Chloro-3-(trifluoromethyl)-2-pyridine carboxylic acid
Formel: C₇H₃ClF₃NO₂
Molecular Weight: 225.56 g/mol
Schmelzpunkt: 136 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD14155643
CAS-Nummer: 1198475-43-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR