Methyl 3-bromo-4-(((trifluoromethyl)sulphonyl)oxy)-5-(trimethylsilyl)benzoate

Lieferant: Apollo Scientific

PC200120-100MG
APOSPC200120-100MGEA 205 EUR
APOSPC200120-100MG
Methyl 3-bromo-4-(((trifluoromethyl)sulphonyl)oxy)-5-(trimethylsilyl)benzoate
Methyl 3-bromo-4-(((trifluoromethyl)sulphonyl)oxy)-5-(trimethylsilyl)benzoate
Formel: C₁₂H₁₄BrF₃O₅SSi
Molecular Weight: 435,286 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD25948970
CAS-Nummer: 1242069-33-5
UN: 1759
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR