2-Chloro-4-(difluoromethyl)-6-methoxypyridine

Lieferant: Apollo Scientific

PC200121-500MG
APOSPC200121-500MGEA 539 EUR
APOSPC200121-500MG
2-Chloro-4-(difluoromethyl)-6-methoxypyridine
2-Chloro-4-(difluoromethyl)-6-methoxypyridine
Formel: C₇H₆ClF₂NO
Lagertemperatur: Raumtemperatur
MDL: MFCD25478549
CAS-Nummer: 1446786-33-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR