2,5-Dichloro-4-(difluoromethyl)pyridine 97%

Lieferant: Apollo Scientific
Achtung

PC200124-1G
APOSPC200124-1GEA 430 EUR
APOSPC200124-1G
2,5-Dichloro-4-(difluoromethyl)pyridine 97%
2,5-Dichloro-4-(difluoromethyl)pyridine
Formel: C₆H₃Cl₂F₂N
Lagertemperatur: Raumtemperatur
MDL: MFCD22199291
CAS-Nummer: 1374659-30-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR