5-(Trifluoromethyl)-1H-pyrazol-3-amine

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Amino-5-(trifluoromethyl)-1H-pyrazole

PC200128-5G
APOSPC200128-5GEA 546 EUR
APOSPC200128-5G
5-(Trifluoromethyl)-1H-pyrazol-3-amine
5-(Trifluoromethyl)-1H-pyrazol-3-amine
Formel: C₄H₄F₃N₃
Molecular Weight: 151,09 g/mol
Schmelzpunkt: 96…99 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09834152
CAS-Nummer: 852443-61-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR