Kalium(3-formylphenyl)trifluoroborat

Lieferant: Apollo Scientific

Synonyme: Potassium trifluoro(3-formylphenyl)borate

PC200133-5G
APOSPC200133-5GEA 74.1 EUR
APOSPC200133-5G
Kalium(3-formylphenyl)trifluoroborat
Kalium(3-formylphenyl)trifluoroborat
Formel: C₇H₅BF₃KO
Molecular Weight: 212.02 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07784378
CAS-Nummer: 871231-44-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR