Potassium-5-formylthiophene-2-trifluoroborate

Lieferant: Apollo Scientific

PC200134-1G PC200134-500MG PC200134-5G
APOSPC200134-1GEA 32.2 EUR
APOSPC200134-1G APOSPC200134-500MG APOSPC200134-5G
Potassium-5-formylthiophene-2-trifluoroborate
Potassium-5-formylthiophene-2-trifluoroborate
Formel: C₅H₃BF₃KOS
Lagertemperatur: Raumtemperatur
MDL: MFCD09800740
CAS-Nummer: 1025113-78-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR