3-(Trifluoromethyl)pyridine-2-carbonitrile 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-(Trifluoromethyl)-2-picolinonitrile, 2-Cyano-3-(trifluoromethyl)pyridine, 3-(Trifluoromethyl)picolinonitrile
PC200149-25G
APOSPC200149-25GEA
104
EUR
APOSPC200149-25G
3-(Trifluoromethyl)pyridine-2-carbonitrile 98%
3-(Trifluoromethyl)pyridine-2-carbonitrile
Formel:
C₇H₃F₃N₂ Molecular Weight: 172.107 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD02091399 CAS-Nummer: 406933-21-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...