3-(Trifluoromethyl)-5-isoxazolamine 95+%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Amino-3-(trifluoromethyl)isoxazole

PC200153-1G PC200153-500MG
APOSPC200153-1GEA 314 EUR
APOSPC200153-1G APOSPC200153-500MG
3-(Trifluoromethyl)-5-isoxazolamine 95+%
3-(Trifluoromethyl)-5-isoxazolamine
Formel: C₄H₃F₃N₂O
Molecular Weight: 152.08 g/mol
Schmelzpunkt: 53…55 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04972407
CAS-Nummer: 108655-63-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR