Ethyl-3-trifluoromethyl-1H-pyrazole-5-carboxylate
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Ethyl 3-(Trifluoromethyl)pyrazole-5-carboxylate
PC200156-1G
APOSPC200156-1GEA
89.9
EUR
APOSPC200156-1G
Ethyl-3-trifluoromethyl-1H-pyrazole-5-carboxylate
Ethyl-3-trifluoromethyl-1H-pyrazole-5-carboxylate
Formel:
C₇H₇F₃N₂O₂ Schmelzpunkt: 93…98 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04111238 CAS-Nummer: 129768-30-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...