6-(Trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylic acid monohydrate
Lieferant: Apollo Scientific
PC200162-1G
APOSPC200162-1GEA
217
EUR
APOSPC200162-1G
6-(Trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylic acid monohydrate
6-(Trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylic acid monohydrate
Formel:
C₉H₅F₃N₂O₂·H₂O Molecular Weight: 248,16 g/mol Schmelzpunkt: 238…239 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09258865 CAS-Nummer: 1018828-69-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...