Kalium(4-(hydroxymethyl)phenyl)trifluoroborat

Lieferant: Apollo Scientific

PC200164-1G PC200164-5G
APOSPC200164-1GEA 0 EUR
APOSPC200164-1G APOSPC200164-5G
Kalium(4-(hydroxymethyl)phenyl)trifluoroborat
Potassium-4-(hydroxymethyl)phenyltrifluoroborate
Formel: C₇H₇BF₃KO
Lagertemperatur: Raumtemperatur
MDL: MFCD08276800
CAS-Nummer: 1015082-78-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR