4-(Trifluormethoxy)phenylboronsäurepinakolester 98%

Lieferant: Apollo Scientific

Synonyme: 4,4,5,5-Tetramethyl-2-(4-(trifluoromethoxy)phenyl)-1,3,2-dioxaborolane

PC200177-25G
APOSPC200177-25GEA 98.6 EUR
APOSPC200177-25G
4-(Trifluormethoxy)phenylboronsäurepinakolester 98%
4-(Trifluormethoxy)phenylboronsäurepinakolester
Formel: C₁₃H₁₆BF₃O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD05863920
CAS-Nummer: 474709-28-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR