Potassium-2-thienyltrifluoroborate
Lieferant: Apollo Scientific
Synonyme:
Potassium 2-Thienyltrifluoroborate
PC200178-1G
PC200178-500MG
PC200178-5G
APOSPC200178-1GEA
102
EUR
APOSPC200178-1G
APOSPC200178-500MG
APOSPC200178-5G
Potassium-2-thienyltrifluoroborate
Potassium-2-thienyltrifluoroborate
Formel:
C₄H₃BF₃KS Molecular Weight: 190.03 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04112719 CAS-Nummer: 906674-55-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...