Kalium(1-naphthyl)trifluoroborat

Lieferant: Apollo Scientific

PC200179-10G PC200179-1G PC200179-5G
APOSPC200179-10GEA 256 EUR
APOSPC200179-10G APOSPC200179-1G APOSPC200179-5G
Kalium(1-naphthyl)trifluoroborat
Kalium(1-naphthyl)trifluoroborat
Formel: C₁₀H₇BF₃K
Lagertemperatur: Raumtemperatur
MDL: MFCD06659936
CAS-Nummer: 166328-07-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR