2,2,2-Trifluoro-1-(1H-pyrrol-3-yl)-1-ethanone

Lieferant: Apollo Scientific

PC200185-1G PC200185-500MG
APOSPC200185-1GEA 386 EUR
APOSPC200185-1G APOSPC200185-500MG
2,2,2-Trifluoro-1-(1H-pyrrol-3-yl)-1-ethanone
2,2,2-Trifluoro-1-(1H-pyrrol-3-yl)-1-ethanone
Formel: C₆H₄F₃NO
Schmelzpunkt: 110…111 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD12924726
CAS-Nummer: 130408-89-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR