3-Iodo-5-(trifluoromethyl)-2-pyridinylamine
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
3-Iodo-5-(trifluoromethyl)pyridin-2-amine, 2-Amino-3-iodo-5-(trifluoromethyl)pyridine
PC200189-5G
APOSPC200189-5GEA
179
EUR
APOSPC200189-5G
3-Iodo-5-(trifluoromethyl)-2-pyridinylamine
3-Iodo-5-(trifluoromethyl)-2-pyridinylamine
Formel:
C₆H₄F₃IN₂ Molecular Weight: 288.01 g/mol Schmelzpunkt: 97…98 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD10697686 CAS-Nummer: 211308-82-6 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...