tert-Butyl-N-(6-chloro-4-fluoro-3-pyridinyl)carbamate > 90%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Amino-2-chloro-4-fluoropyridine, 5-BOC protected

PC200192-1G
APOSPC200192-1GEA 473 EUR
APOSPC200192-1G
tert-Butyl-N-(6-chloro-4-fluoro-3-pyridinyl)carbamate > 90%
tert-Butyl-N-(6-chloro-4-fluoro-3-pyridinyl)carbamate
Formel: C₁₀H₁₂ClFN₂O₂
Molecular Weight: 246,67 g/mol
Schmelzpunkt: 90…91 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07368891
CAS-Nummer: 915307-78-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR