3-Fluorophenylboronic acid pinacol ester

Lieferant: Apollo Scientific
Achtung

PC200195-10G PC200195-1G PC200195-5G
APOSPC200195-10GEA 43.9 EUR
APOSPC200195-10G APOSPC200195-1G APOSPC200195-5G
3-Fluorophenylboronic acid pinacol ester
3-Fluorophenylboronic acid pinacol ester
Formel: C₁₂H₁₆BFO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD18375236
CAS-Nummer: 936618-92-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR