3-Chloro-5-(trifluoromethyl)-2-pyridinecarbohydrazide
Lieferant: Apollo Scientific
|
Achtung
|
PC200201-1G
PC200201-500MG
APOSPC200201-1GEA
538
EUR
APOSPC200201-1G
APOSPC200201-500MG
3-Chloro-5-(trifluoromethyl)-2-pyridinecarbohydrazide
3-Chloro-5-(trifluoromethyl)-2-pyridinecarbohydrazide
Formel:
C₇H₅ClF₃N₃O Schmelzpunkt: 105…107 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD10758083 CAS-Nummer: 1033463-30-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...