1-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]azepane

Lieferant: Apollo Scientific

PC200216-1G PC200216-500MG PC200216-5G
APOSPC200216-1GEA 132 EUR
APOSPC200216-1G APOSPC200216-500MG APOSPC200216-5G
1-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]azepane
1-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]azepane
Formel: C₁₂H₁₄ClF₃N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD13561841
CAS-Nummer: 1219967-95-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR