3-Fluoro-5-(trifluoromethyl)pyridine-2-carbonitrile

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Fluoro-5-(trifluoromethyl)picolinonitrile

PC200219-1G PC200219-5G
APOSPC200219-1GEA 27.7 EUR
APOSPC200219-1G APOSPC200219-5G
3-Fluoro-5-(trifluoromethyl)pyridine-2-carbonitrile
3-Fluoro-5-(trifluoromethyl)pyridine-2-carbonitrile
Formel: C₇H₂F₄N₂
Molecular Weight: 190.1 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11848039
CAS-Nummer: 80194-71-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR