5-Fluoroimidazo[1,2-a]pyridine-2-carbaldehyde
Lieferant: Apollo Scientific
|
Achtung
|
PC200220-500MG
APOSPC200220-500MGEA
419
EUR
APOSPC200220-500MG
5-Fluoroimidazo[1,2-a]pyridine-2-carbaldehyde
5-Fluoroimidazo[1,2-a]pyridine-2-carbaldehyde
Formel:
C₈H₅FN₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD11656369 CAS-Nummer: 878197-67-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...