6-Fluorimidazo[1,2-a]pyridin
Lieferant: Apollo Scientific
|
Achtung
|
PC200221-1G
PC200221-10G
PC200221-5G
APOSPC200221-1GEA
79.9
EUR
APOSPC200221-1G
APOSPC200221-10G
APOSPC200221-5G
6-Fluorimidazo[1,2-a]pyridin
6-Fluorimidazo[1,2-a]pyridin
Formel:
C₇H₅FN₂ Schmelzpunkt: 70…71 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09994632 CAS-Nummer: 139022-27-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...