[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]methanol

Lieferant: Apollo Scientific
Achtung

Synonyme: (3-Chloro-5-(trifluoromethyl)pyridin-2-yl)methanol, 3-Chloro-2-(hydroxymethyl)-5-(trifluoromethyl)pyridine

PC200233-250MG
APOSPC200233-250MGEA 42.2 EUR
APOSPC200233-250MG
[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]methanol
[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]methanol
Formel: C₇H₅ClF₃NO
Molecular Weight: 211,57 g/mol
Schmelzpunkt: 36…38 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD11100207
CAS-Nummer: 1033463-31-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR