2-Bromo-6-(trifluoromethyl)nicotinic acid

Lieferant: Apollo Scientific
Achtung

PC200239-1G
APOSPC200239-1GEA 415 EUR
APOSPC200239-1G
2-Bromo-6-(trifluoromethyl)nicotinic acid
2-Bromo-6-(trifluoromethyl)nicotinic acid
Formel: C₇H₃BrF₃NO₂
Molecular Weight: 270.01 g/mol
Schmelzpunkt: 130…132 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09909645
CAS-Nummer: 749875-07-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR