4-[3-(Trifluoromethyl)-2-pyridinyl]benzenecarbaldehyde

Lieferant: Apollo Scientific

PC200241-1G PC200241-500MG PC200241-5G
APOSPC200241-1GEA 0 EUR
APOSPC200241-1G APOSPC200241-500MG APOSPC200241-5G
4-[3-(Trifluoromethyl)-2-pyridinyl]benzenecarbaldehyde
4-[3-(Trifluoromethyl)-2-pyridinyl]benzenecarbaldehyde
Formel: C₁₃H₈F₃NO
Schmelzpunkt: 67…68 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD10758064
CAS-Nummer: 847446-85-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR