tert-Butyl-3-(trifluoromethyl)-1H,4H,5H,6H,7H-pyrazolo[4,3-c]pyridine-5-carboxylate

Lieferant: Apollo Scientific
Achtung

Synonyme: 4,5,6,7-Tetrahydro-3-(trifluoromethyl)-1H-pyrazolo[4,3-c]pyridine, N5-BOC protected, tert-Butyl 3-(trifluoromethyl)-6,7-dihydro-1H-pyrazolo[4,3-c]pyridine-5(4H)-carboxylate

PC200251-500MG
APOSPC200251-500MGEA 242 EUR
APOSPC200251-500MG
tert-Butyl-3-(trifluoromethyl)-1H,4H,5H,6H,7H-pyrazolo[4,3-c]pyridine-5-carboxylate
tert-Butyl-3-(trifluoromethyl)-1H,4H,5H,6H,7H-pyrazolo[4,3-c]pyridine-5-carboxylate
Formel: C₁₂H₁₆F₃N₃O₂
Molecular Weight: 291,28 g/mol
Schmelzpunkt: 170…172 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD11044975
CAS-Nummer: 1022931-73-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR