3-Fluoro-5-(trifluoromethyl)pyridine-2-carboxamide 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC200268-500MG
APOSPC200268-500MGEA
288
EUR
APOSPC200268-500MG
3-Fluoro-5-(trifluoromethyl)pyridine-2-carboxamide 95%
3-Fluoro-5-(trifluoromethyl)pyridine-2-carboxamide
Formel:
C₇H₄F₄N₂O Schmelzpunkt: 168…169 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD16038708 CAS-Nummer: 852062-16-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...