Methyl-2-[3-chloro-5-(trifluoromethyl)-2-pyridinyl]acetate

Lieferant: Apollo Scientific
Achtung

PC200274-1G
APOSPC200274-1GEA 317 EUR
APOSPC200274-1G
Methyl-2-[3-chloro-5-(trifluoromethyl)-2-pyridinyl]acetate
Methyl-2-[3-chloro-5-(trifluoromethyl)-2-pyridinyl]acetate
Formel: C₉H₇ClF₃NO₂
Molecular Weight: 253,61 g/mol
Schmelzpunkt: 38…40 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07366545
CAS-Nummer: 885949-63-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR