Ethyl 2-cyano-5-(trifluoromethyl)nicotinate
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Ethyl 2-cyano-5-(trifluoromethyl)nicotinate
PC200275-500MG
APOSPC200275-500MGEA
300
EUR
APOSPC200275-500MG
Ethyl 2-cyano-5-(trifluoromethyl)nicotinate
Ethyl-2-cyano-5-(trifluoromethyl)nicotinate
Formel:
C₁₀H₇F₃N₂O₂ Molecular Weight: 244.17 g/mol Schmelzpunkt: 55…57 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD13193103 CAS-Nummer: 1192263-82-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...