2,3-Dichloro-5-(trifluoromethyl)isonicotinic acid
Lieferant: Apollo Scientific
PC200277-1G
APOSPC200277-1GEA
317
EUR
APOSPC200277-1G
2,3-Dichloro-5-(trifluoromethyl)isonicotinic acid
2,3-Dichloro-5-(trifluoromethyl)isonicotinic acid
Formel:
C₇H₂Cl₂F₃NO₂ Schmelzpunkt: 192…193 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD12025872 CAS-Nummer: 1135283-33-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...