2-{3-[2-Amino-5-(trifluoromethyl)-3-pyridinyl]-2-propynyl}-1H-isoindole-1,3(2H)-dione

Lieferant: Apollo Scientific

PC200309-500MG
APOSPC200309-500MGEA 524 EUR
APOSPC200309-500MG
2-{3-[2-Amino-5-(trifluoromethyl)-3-pyridinyl]-2-propynyl}-1H-isoindole-1,3(2H)-dione
2-{3-[2-Amino-5-(trifluoromethyl)-3-pyridinyl]-2-propynyl}-1H-isoindole-1,3(2H)-dione
Formel: C₁₇H₁₀F₃N₃O₂
Schmelzpunkt: 186…188 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD14584841
CAS-Nummer: 1221792-28-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR