5-(Trifluoromethyl)-1H-pyrrolo[2,3-b]pyridine-3-carbaldehyde

Lieferant: Apollo Scientific

PC200311-500MG
APOSPC200311-500MGEA 386 EUR
APOSPC200311-500MG
5-(Trifluoromethyl)-1H-pyrrolo[2,3-b]pyridine-3-carbaldehyde
5-(Trifluoromethyl)-1H-pyrrolo[2,3-b]pyridine-3-carbaldehyde
Formel: C₉H₅F₃N₂O
Molecular Weight: 214,15 g/mol
Schmelzpunkt: 211…212 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD11841013
CAS-Nummer: 1135283-53-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR