4-Chloro-2-(trifluoromethyl)-8-quinolinol

Lieferant: Apollo Scientific

PC200317-500MG
APOSPC200317-500MGEA 0 EUR
APOSPC200317-500MG
4-Chloro-2-(trifluoromethyl)-8-quinolinol
4-Chloro-2-(trifluoromethyl)-8-quinolinol
Formel: C₁₀H₅ClF₃NO
Lagertemperatur: Raumtemperatur
MDL: MFCD08056688
CAS-Nummer: 886362-20-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR