8-Methyl-2-(trifluoromethyl)imidazo[1,2-a]pyridine-3-carboxylic acid

Lieferant: Apollo Scientific

PC200323-500MG
APOSPC200323-500MGEA 292 EUR
APOSPC200323-500MG
8-Methyl-2-(trifluoromethyl)imidazo[1,2-a]pyridine-3-carboxylic acid
8-Methyl-2-(trifluoromethyl)imidazo[1,2-a]pyridine-3-carboxylic acid
Formel: C₁₀H₇F₃N₂O₂
Molecular Weight: 244,17 g/mol
Schmelzpunkt: 196…197 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03407573
CAS-Nummer: 874776-53-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR